Learn More
Medchemexpress LLC Coenzyme Q8 (ubiquinone 8) | 2394-68-5 | 99.0% | 727.11 | 1 MG
Supplier: Medchemexpress LLC HYN96801MG
Coenzyme Q8 (Ubiquinone 8) is an isoprenoid quinone that mediates electron transfer within the aerobic respiratory chain and reduces oxidative stress. It maintains bacterial respiratory function, enhances host resistance to bacterial infection, stimulates macrophage phagocytosis, and increases antibody-producing cells. It can be used in infectious disease research.
- Enhances phagocytosis of sheep erythrocytes by peritoneal macrophages
- Provides complete protection against otherwise lethal infections with gram-negative and gram-positive bacteria
- Enhances bacterial clearance from blood
- Stimulates macrophage phagocytosis
- Increases antibody-producing cells in spleens
- Storage: Powder at -20°C for 3 years; In solvent at -80°C for 6 months, -20°C for 1 month