Learn More
Medchemexpress LLC Cofrogliptin | 1844874-26-5 | 99.29% | 466.43 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY147257100MG
Encompass_Preferred
Cofrogliptin (HSK7653), a tetrahydropyran derivative, is a potent oral dipeptidyl aminopeptidase 4 (DPP-4) inhibitor with long-acting antidiabetic efficacy. It has great potential for type 2 diabetes mellitus (T2DM).
  • Potent oral dipeptidyl aminopeptidase 4 (DPP-4) inhibitor
  • Long-acting antidiabetic efficacy
  • Potential for treating type 2 diabetes mellitus (T2DM)
  • DPP-4 inhibitory activity with an IC₅₀ value of 4.18 nM
  • Exhibits extremely long half-lives
  • Low rate of drug concentration reduction after oral administration
Catalog No. 50-003-61979