Learn More
Selleck Chemical LLC COH-SR4-E0795-5MG
SDP
Supplier:  Selleck Chemical LLC E07955MG
Encompass_Preferred
COH-SR4 suppresses adipogenesis in 3T3-L1 cells through indirect activation of AMPK and downstream modulation of the Mammalian target of rapamycin complex 1 (mTORC1) signaling pathway which blocks important regulators involved in protein synthesis cell cycle progression and expression of key transcription factors and their target adipogenic genes involved in lipid synthesis
Catalog No. 50-313-7882