Learn More
MSC Value Collection Coiled Spring Pin, 1070/1090 Alloy Steel, 5/32 in. Diameter, 3/4 in. Length, 1 ea.
Supplier: MSC 67502401
These coiled spring pins, also referred to as spiral pins, are engineered with multiple turns of alloy steel to provide superior shock absorbance and added strength. The coiled design allows for even distribution of stress across the pin, making them ideal for various industrial fastening applications requiring robust performance.
- Manufactured from 1070/1090 alloy steel
- Plain finish
- Diameter of 5/32 in.
- Overall length of 3/4 in.
- Designed for even stress distribution
- Provides enhanced shock absorbance