Learn More
ABclonal Technology COL4A3BP Rabbit pAb
SDP
Supplier:  ABclonal Technology A153761000UL
Encompass_Preferred
This gene encodes a kinase that specifically phosphorylates the N-terminal region of the non-collagenous domain of the alpha 3 chain of type IV collagen, known as the Goodpasture antigen. Goodpasture disease is the result of an autoimmune response directed at this antigen. One isoform of this protein is also involved in ceramide intracellular transport. Three transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-05865