Learn More
FUJIFILM BIOSCIENCES INC COLCEMID SOLUTION 1X10ML
Supplier:  FUJIFILM BIOSCIENCES INC 931110ML
Encompass
Colcemid Solution arrests mitosis in both plant and animal cells, preventing spindle formation during metaphase. This formulation is 10 �g/ml, prepared in Hanks' Balanced Salts, packaged with SEPTUM CAPS, and is pipette and syringe accessible. 10ml.
Catalog No. NC9661266