Learn More
Apexbio Technology LLC Colchicine 64-86-8 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A33245.1
Encompass_Preferred
Colchicine (CAS 64-86-8) is a small-molecule inhibitor targeting tubulin polymerization It binds directly to tubulin monomers disrupting their polymerization into microtubules (IC50 3 2 M) By preventing microtubule assembly colchicine interferes with spindle formation arresting cells in mitosis at metaphase thereby functioning as a mitotic inhibitor Additionally colchicine demonstrates anti-inflammatory activity by suppressing neutrophil motility and function reducing urate crystal deposition and has been effectively used in animal models of gouty arthritis It is widely utilized in biomedical research to study microtubule dynamics mitosis control and inflammatory pathways
Catalog No. 50-000-62830