Learn More
Selleck Chemical LLC Colchicine S2284-10mM/1mL
SDP
Supplier:  Selleck Chemical LLC S228410MM/1ML
Encompass_Preferred
Colchicine is a microtubule polymerization inhibitor with an IC50 of 3 nM
Catalog No. 50-311-2650