Learn More
AdipoGen Colchicine,Chemical. CAS: 64-86-8
SDP
Supplier:  AdipoGen AGCN20048M500
Encompass_Preferred
Colchicine, AdipoGen Life Sciences. Chemical. CAS: 64-86-8. Formula: C22H25NO6. MW: 399.4. Isolated from Colchicum autumnale. Anti-cancer compound. Microtubule assembly inhibitor. Depolymerizes microtubules and limits microtubule formation (inactivates spindle fibre formation). Inhibits mitosis during cell division at metaphase by inhibiting spindle formation. Anti-inflammatory compound. Suppresses monosodium urate crystal-induced NLRP3/NALP3 inflammasome-driven caspase-1 activation, IL-1beta processing and release, and L-selectin expression on neutrophils at micromolar concentrations. Blocks the release of a crystal-derived chemotactic factor from neutrophil lysosomes, blocks neutrophil adhesion to, and inhibits monosodium urate crystal-induced production of superoxide anions from neutrophils at nanomolar concentrations. Drug used in treatment of gout, familial Mediterranean fever, pericarditis and Behect's disease. Investigated for its anti-cancer activity.
Catalog No. 50-114-6238