Learn More
Sigma Aldrich Fine Chemicals Biosciences Colchicine 95 HPLC powder5G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences C97545G
Encompass
Colchicine is a secondary metabolite which has anti-inflammatory properties. It is used to treat gout crystal-induced joint inflammation familial Mediterranean fever BehA§et disease vasculitides andnbsppericarditis. Colchicine inhibits cell division intracellular vesicle motility secretion of cytokines andnbspchemokines. It is implicated innbspleukocyte diapedesis and lysosomal degranulation. Colchicine also blocks fibroblast proliferation and collagen transport to extracellular space.
Catalog No. 50-290-3959