Learn More
Medchemexpress LLC Colchicine-d3 | 1217625-62-1 | 99.2% | 402.46 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY16569S11MG
Encompass_Preferred
Colchicine-d3 is the deuterium labeled Colchicine. It acts as a tubulin inhibitor and a microtubule disrupting agent, effectively inhibiting microtubule polymerization with an IC50 of 3 nM. This compound also functions as a competitive antagonist of the α3 glycine receptors (GlyRs).
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Inhibits microtubule polymerization
  • Competitive antagonist of α3 glycine receptors
Catalog No. 50-003-91793