Learn More
Medchemexpress LLC Colchicine-d6 | 1217651-73-4 | 99.4% | 405.47 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY16569S1MG
Encompass_Preferred
Colchicine-d6 is a deuterium-labeled version of Colchicine. Colchicine acts as a tubulin inhibitor and a microtubule disrupting agent, inhibiting microtubule polymerization with an IC50 of 3 nM. It also functions as a competitive antagonist of the α3 glycine receptors (GlyRs).
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-91805