Learn More
Sigma Aldrich Fine Chemicals Biosciences Colchicine suitable for pl5G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences C39155G
Encompass
Colchicine is a tricyclic alkaloid that can stay in circulation for 20 to 40 hours post intake. Leucocytes harbour maximum colchicine. It contains three hexameric rings termed A B and C.
Catalog No. 50-291-4264