Learn More
Electron Microscopy Sciences Cold Embedding Molds, 1.25" diameter
SDP
Supplier:  Electron Microscopy Sciences 5048122
Encompass_Preferred
These 2 part molds may be used for all cold embedding epoxies. Comes with a detached cap for easy removal. The molds come in 3 different sizes to accommodate all of the samples you may have. The molds are made from HDPE and we recommend the use of mold release spray prior to use.
Diameter: 1.25"
Catalog No. 50-192-6833