Learn More
Electron Microscopy Sciences EMS Vibrating Microtome Cold Light Source
SDP
Supplier:  Electron Microscopy Sciences 700021
Encompass_Preferred
Optional cold light source for EMS7000smz Vibrating Microtome povides direct the light where needed with Twin 500mm fiber optic light guides. Light Intensity is adjustable by a rotary dial knob. 20W/12V Halogen lamp.
Catalog No. 50-949-850