Learn More
Cable Ties And More COLD WEATHER ZIP TIES 7" CS100
SDP
Supplier:  Cable Ties And More CTCW7
Encompass_Preferred
Specialized Nylon Cable Ties Suited For -40º F Cold Temperatures.100 Pcs Per Pack. 7" long.50 Lbs Tensile Strength.Tested For Application Performance at -20°C/-4°F Material Rated At -40°C/-40°F Per ISO180 ROHS Compliant
Catalog No. NC1917300