Learn More
Shockwatch Div Mri ColdMark Descending Temperature Indicator 0C/32F
SDP
Supplier:  Shockwatch Div Mri CM 02/32
Encompass_Preferred
ColdMark temperature indicator monitors temperatures during shipping and storage, indicating if goods experience unacceptable conditions. These reliable temperature indicators provide visual evidence of exposure to unacceptably low temperature levels, allowing you to determine if product quality has been compromised. Temperature threshold: 0C/32F. Accuracy +/-1C. Shelf life 2 years. Single-use.
Catalog No. NC2228394