Learn More
Medchemexpress LLC Colestilan | 95522-45-5 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11293950MG
Encompass_Preferred
Colestilan is a non-absorbed, non-calcium-based phosphate binder and a non-metallic, anion exchange resin. It is orally active and can be used for hypercholesterolaemia research. This product is for research use only.
  • Decreases cholesterol levels by bile acid adsorption
  • Binds phosphate in vitro
  • Dose-dependently decreases blood phosphate and urinary phosphate excretion in rats
  • Improves hyperphosphataemia in rats with renal failure
Catalog No. 50-004-39873