Learn More
Medchemexpress LLC Colestilan (MCI-196) | 95522-45-5 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY112939
Encompass_Preferred
Colestilan (MCI-196) is a non-absorbed, non-calcium-based phosphate binder and is also a non-metallic, anion exchange resin. Colestilan is orally active and can be used for hypercholesterolaemia research.
  • Non-absorbed.
  • Non-calcium-based phosphate binder.
  • Non-metallic, anion exchange resin.
  • Orally active.
  • Can be used for hypercholesterolaemia research.
  • Decreases cholesterol levels by bile acid adsorption through the gastrointestinal tract.
  • Binds phosphate in vitro.
  • Decreases blood phosphate and urinary phosphorus excretion in rats.
  • Able to improve hyperphosphataemia in rats with renal failure.
Catalog No. 50-004-39419