Learn More
Medchemexpress LLC Colestilan (MCI-196) | 95522-45-5 | 98.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY112939100MG
Encompass_Preferred
Colestilan (MCI-196) is a non-absorbed, non-calcium-based phosphate binder and a non-metallic, anion exchange resin. It is orally active and can be used for hypercholesterolaemia research. This compound decreases cholesterol levels by bile acid adsorption through the gastrointestinal tract.
  • Non-absorbed, non-calcium-based phosphate binder
  • Non-metallic, anion exchange resin
  • Orally active
  • Decreases cholesterol levels by bile acid adsorption
  • Binds 2.52 mmol of phosphate at 2-4 mM phosphate concentration and pH = 7.2 in vitro
  • Decreases blood phosphate and urinary phosphorus excretion in rats in vivo
  • Improves hyperphosphataemia in rats with renal failure
Catalog No. 50-004-39751