Learn More
Medchemexpress LLC Colfosceril miristate | 18194-24-6 | 99.96% | 677.93 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY109541
Encompass_Preferred
Colfosceril miristate (DMPC) is a synthetic phospholipid with antiproliferative effects on various tumors. It is widely used in studies of lipid monolayers, bilayers, and drug delivery systems.
  • Utilized in studies of lipid monolayers, bilayers
  • Supports drug delivery research
  • Exhibits cytotoxicity on certain tumor cell lines
  • Enhances nanoparticle entry into PC-12 cells
  • Promotes diffusion of SPIONs in rat brain
Catalog No. 50-003-47584