Learn More
Medchemexpress LLC Colfosceril miristate (DMPC) | 18194-24-6 | 100.0% | 677.93 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY109541S51MG
Encompass_Preferred
Colfosceril miristate (DMPC) is a synthetic phospholipid that exhibits antiproliferative effects on various tumors and is utilized in studies of lipid monolayers, bilayers, and drug delivery. It can stabilize Carmofur at neutral pH, shows cytotoxicity on several tumor cell lines (SHSY-5Y, MDA-MB-435s, and HT-1080), and enhances the ability of nanoparticles to enter PC-12 cells. In vivo, it promotes the diffusion of SPIONs from the substantia nigra to nearby brain regions in SD rats.
  • Synthetic phospholipid
  • Exhibits antiproliferative effects on tumors
  • Used in studies of lipid monolayers, bilayers, and drug delivery
  • Stabilizes Carmofur at neutral pH
  • Shows cytotoxicity on various tumor cell lines
  • Enhances nanoparticle entry into cells
  • Promotes diffusion of SPIONs in brain regions
Catalog No. 50-003-44893