Learn More
World Precision Instrument Colibri Suturing Forceps with Tying Platform, 7.5 cm
SDP
Supplier:  World Precision Instrument WP1520
Encompass
  • 7.5 cm (3") long
  • Tying platform
  • 1x2 140° Teeth
  • Titanium
Catalog No. 50-253-8339