Learn More
IDEXX REFERENCE LABORATORIES COLILERT-18 Test, 200 Test Pack
Supplier:  IDEXX REFERENCE LABORATORIES WP200I18
Encompass
This supplier will only sell to Pharmaceutical and Manufacturing companies. They will not ship to Universities or Hospitals. Simultaneously detects both total coliforms and Escherichia coli in water, or fecal coliforms in wastewater, giving you results in 18 hours. Read afternoon samples the next morningbefore the next days samples arrive. Lift boil water alerts in 18 hours. Provide results in record time for real estate, new well, and new construction samples.
Catalog No. NC1620794