Learn More
Medchemexpress LLC Colivelin | 867021-83-8 | 99.9% | 2645.10 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP10615MG
Encompass_Preferred
Colivelin is a brain penetrant neuroprotective peptide and a potent activator of STAT3. It suppresses neuronal death by activating STAT3 in vitro. Colivelin demonstrates long-term beneficial effects against neurotoxicity, Aβ deposition, neuronal apoptosis, and synaptic plasticity deficits in neurodegenerative disease. It has potential for treating Alzheimer's disease and ischemic brain injury.
  • Suppresses neuronal death by activating STAT3 in vitro
  • Demonstrates long-term beneficial effects against neurotoxicity, Aβ deposition, neuronal apoptosis, and synaptic plasticity deficits
  • Potential for treating Alzheimer's disease and ischemic brain injury
Catalog No. 50-004-25582