Learn More
Medchemexpress LLC Collagen I alpha 2 Antibody (YA3695) | 10 UL
SDP
Supplier:  Medchemexpress LLC HYP8399810UL
Encompass_Preferred
Collagen I alpha 2 Antibody (YA3695) is a Mouse-derived, non-conjugated IgG1 monoclonal antibody targeting Collagen I alpha 2. Intended for research use only, this antibody is supplied in PBS with 0.05% sodium azide and exhibits reactivity with human, mouse, rat, and monkey species.
  • Mouse-derived IgG1 monoclonal antibody
  • Targets Collagen I alpha 2
  • For research use only
  • Supplied in PBS with 0.05% sodium azide
  • Reacts with human, mouse, rat, and monkey species
Catalog No. 50-004-44763