Learn More
Genetex Inc Collagen IV Monoclonal Antibody [MSVA-704R] HistoMAX™ (GTX04434) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04434
Encompass_Preferred
The Collagen IV antibody [MSVA-704R] HistoMAX™ (GTX04434) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human lamina muscularis mucosae from stomach tissue using GTX04434 Collagen IV antibody [MSVA-704R] HistoMAX. In the intestine collagen IV immunostaining involves basement membranes vessels of all sizes and smooth muscle cells of the lamina muscularis mucosae.
Catalog No. NC3666942