Learn More
Ibidi USA Collagen Type I, Rat Tail (10 mg/ml , 1 x 5 ml Bottle)
SDP
Supplier:  Ibidi USA 50204
Encompass
Type I Collagen is a major structural component of the extracellular matrix (ECM). This fibrous protein is often used in three-dimensional (3D) collagen gels that simulate the in vivo cell environment better than the traditional 2D systems. Collagen I can also be used for coating surfaces, as it can form thin layers for culturing cells. ibidi’s Type I Collagen is a state-of-the-art, purified protein extracted from rat tail tendons without pepsinization. It is specially engineered for ibidi’s cell-based assays (e.g., chemotaxis assays in 3D gels, or 2D coating of lab cultureware). sterile, 10 mg/ml, 1 x 5 ml trial pack
Catalog No. 50-114-9016