Learn More
Sigma Aldrich Fine Chemicals Biosciences Collagenase from Clostridium histolyticum, 9001-12-1, MFCD00130830, 50 mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences C972250MG
Encompass
Lyophilized powder (from sterile-filtered solution), 0.5-5.0 FALGPA units/mg solid, cell culture tested, Synonym: Clostridiopeptidase A. This product is suitable for the disaggregation of human tumor, mouse kidney, human adult and fetal brain, lung and many other epithelia tissues. It has also been shown to be effective in liver and kidney perfusion studies, digestion of pancreas, isolation of nonparenchymal rat liver cells and hepatocyte preparation.
Catalog No. NC1308897