Learn More
Sigma Aldrich Fine Chemicals Biosciences Collagenase A from Clostri
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 11088793001
Encompass
Collagenase is a protease which cleaves the triple-helical protein called collagen. Clostridium histolyticum produces six types of collagenases namely ³ ´ µ and». These collagenases are active against connective tissue and show more reactivity towards gelatin than collagen. Collagenase obtained from Clostridium histolyticum has a very strong activity as it digests collagen from both ends at temperatures as low as 4-10 C.
Catalog No. 50-290-8200