Learn More
Worthington Biochemical Corporation COLLAGENASE CLSAFA FI 50MG
SDP
Supplier:  Worthington Biochemical Corporation LS004118
Encompass
Collagenase, Animal Origin-Free (Code: CLSAFA) which is filtered through a 0.22 micron membrane and lyophilized in vials. 2593-92-3
Catalog No. 50-592-431