Learn More
Sigma Aldrich Fine Chemicals Biosciences Collagenase/Dispase, 100 mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 10269638001
Encompass
Collagenase/Dispase is used for the preparation of cells from a wide variety of different tissues and organs.
Catalog No. NC0953723