Learn More
Sigma Aldrich Fine Chemicals Biosciences Collagenase from Clostridium histolyticum release of physiologically active rat epididymal adipocytes tested, Type II, 0.5-5.0 FALGPA units/mg solid, >=125 CDU/mg solid | 9001-12-1 | MFCD00130830 | 1G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences C68851G
Encompass
Collagenase from Clostridium histolyticum release of physiologically active rat epididymal adipocytes tested, Type II, 0.5-5.0 FALGPA units/mg solid, >=125 CDU/mg solid | 9001-12-1 | MFCD00130830 | 1G
Catalog No. 50-178-0967