Learn More
Sigma Aldrich Fine Chemicals Biosciences Collagenase H from Clostri
Supplier:  Sigma Aldrich Fine Chemicals Biosciences COLLHRO
Encompass
Collagenase H is an enzyme mixture prepared from Clostridium histolyticum cultures by filtration ammonium sulfate precipitation dialysis and lyophilization.
Catalog No. 50-290-7744