Learn More
Worthington Biochemical Corporation COLLAGENASE IV /5GM
SDP
Supplier:  Worthington Biochemical Corporation LS004189
Encompass
Prepared to contain lower tryptic activity levels to limit damage to membrane proteins and receptors but with normal to above normal collagenase activity. Suggested for pancreatic islet primary isolation. A dialyzed, lyophilized powder.
Catalog No. NC0287247