Learn More
Worthington Biochemical Corporation Collagenase, Type 3, 5g
SDP
Supplier:  Worthington Biochemical Corporation LS004183
Encompass
Lower in secondary proteolytic contaminant activities but with typical collagenase activity. Suggested for mammary primary cell isolation. A dialyzed, lyophilized powder. Store at 2-8°C.
Catalog No. NC0021929