Learn More
Worthington Biochemical Corporation Collagenase Vial, NCIS, 5 vials
SDP
Supplier:  Worthington Biochemical Corporation LK003245
Encompass
A component of the NCIS kit. This material is 0.22 micron membrane filtered and lyophilized in autoclaved vials. A vial reconstituted with 5 ml of HBSS or equivalent yields a solution of 300 units/ml of collagenase, Code: CLSPA. Suitable for cell isolation and culture applications. Store at 2-8°C.
Catalog No. NC9300431