Learn More
Fritsch Milling And Sizing Inc Collecting Vessel 60 Litres with Hinge + Filter Hose, P-15
SDP
Supplier:  Fritsch Milling And Sizing Inc 15.4560.00
Encompass
For grinding large quantities this collecting vessel with a capacity of 60 litres and filter hose is recommended for PULVERISETTE 15.
For smaller samples a collecting vessel 3.5 litres is recommended order no. 15.4400.00
Catalog No. 50-153-4010