Learn More
Peak Scientific Instruments Ltd Collision Gas Add-On Produces 200cc/min of 99.999% Nitrogen
SDP
Supplier:  Peak Scientific Instruments Ltd 3304886
Encompass_Preferred
Can be added to PEAK gas generator already working in the lab
Ultra High Purity – 99.999% Nitrogen, at 200cc/min.*
Removes unsafe and environmentally unfriendly cylinders from the lab
On-screen performance and alerts for generator status indication
No risk of running out of gas in the middle of analysis
Compatible with the current Genius XE35, XE70, Horizen 24, Infinity XE, and Solaris XE systems
2-year comprehensive manufacturer’s warranty**
*Nitrogen generator providing main gas source to an instrument. (Genius XE35/70, Horizen 24, Infinity XE and Solaris XE systems)
Catalog No. 50-008-04199