Learn More
LIFE TECH (BID ECHO ONLY) Colloidal Blue Staining Kit
Supplier:  LIFE TECH (BID ECHO ONLY) LC6025
Encompass
The Colloidal Blue Staining Kit allows detection of nanogram levels of protein with minimal effort and requires only water to destain. The Colloidal Blue stain uses colloidal chemistry that reduces free dye in solution and improves the protein-to-dye binding ratio. Samples are intensely stained and visible within 3 hours, background staining is virtually eliminated by destaining overnight with water.
  • Easy-to-use, mass spectrometry compatible reagents
  • Great densitometry results
Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
Catalog No. NC3859428