Learn More
Scytek Laboratories Colloidal Iron Stain Kit: Large
Supplier: Scytek Laboratories CIK1
The Colloidal Iron Stain Kit is designed for the histological visualization of acid mucopolysaccharides.Acid Mucopolysaccharides: Bright BlueCollagen: Shades of RedKit contents: AAK500, Acetic Acid Solution (12%), 500 ml.HQA125, Hydrochloric Acid Solution (1N), 125 ml.PFD125, Potassium Ferrocyanide Solution (3%), 125 ml.CIS125, Colloidal Iron Stock Solution, 125 ml.VGS125, Van Gieson’s Solution, 125 ml.