Learn More
Sielc Technologies Inc Primesep 100 column, 100 A, 4.6x50mm, 3um
Supplier:  Sielc Technologies Inc 10046.050.0310
Encompass
Primesep 100 is a reverse-phase analytical column with embedded acidic ion-pairing groups. This is the most versatile column of the entire Primesep family for separation of a broad range of compounds. Improves retention of basic compounds by cation-exchange mechanism. Separates acids by ion-exclusion mechanism. Retains neutral compounds by reverse-phase mechanism. All mobile phases are LC-MS and preparative chromatography compatible.
Catalog No. NC1770568