Learn More
HANNA INSTRUMENTS INC Color of Water Photometer
SDP
Supplier:  HANNA INSTRUMENTS INC HI97727
Encompass
The HI97727 Color of Water Photometer combines accuracy and ease of use in a simple, portable design. The advanced optical system provides lab-quality accuracy while its user-friendly design is easy for any user making it the perfect photometer for your water quality testing needs. The HI97727 meter measures color of water samples from 0 to 500 PCU.No warm up time needed before taking a measurement.Tutorial mode for easy step-by-step instructions.CAL CheckTM for performance verification and calibration.
Catalog No. NC1872441