Learn More
TARGETMOL CHEMICALS INC Columbamine 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T5S080310MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Columbamine (Columbamin) shows strong activity with IC5 48.1 uM.Columbamine exerts anti-proliferative and anti-vasculogenic effects on metastatic human osteosarcoma U2OS cells with low toxicity. Purity 99.16%
Catalog No. 50-287-6596