Learn More
Cayman Chemical Columbamn 10mg
SDP
Supplier:  Cayman Chemical 3503210 MG
Encompass_Preferred
An alkaloid with diverse biological activities; a metabolite of berberine; inhibits CYP3A4 (IC50 = 30.6 µM); active against P. falciparum (IC50 = 1.92 µM); decreases glucose consumption of, and triglyceride levels in, HepG2 cells at 3 and 15 µM, respectively; reduces xylene-induced EAr edema and acetic acid-induced writhing in mice at 56 mg/kg
Catalog No. 50-291-0669