Learn More
Medchemexpress LLC (+)-Columbianetin acetate | 23180-65-6 | 288.30 | 20 MG
SDP
Supplier:  Medchemexpress LLC HYN0363A20MG
Encompass_Preferred
(S)-Columbianetin acetate is an isomer of Columbianetin. Columbianetin is identified as a phytoalexin, which is a substance associated with the resistance of celery (Apium graveolens) to pathogens during storage. This compound has demonstrated significant anti-fungal and anti-inflammatory activities.
  • Associated with celery's resistance to pathogens
  • Exhibits excellent anti-fungal activity
  • Exhibits anti-inflammatory activity
Catalog No. 50-003-88684