Learn More
Waters Corp Waters Column, Glycan BEH C18 AX, 1.7 μm, 2.1 x 150 mm
Supplier:  Waters Corp 186009760
Encompass_Preferred
These columns provide charge-based separation and supreme resolution for acidic glycans using mixed-mode, anion exchange RP technology. They are recommended for UPLC or UHPLC systems, especially for LC-MS methods, and incorporate MaxPeak High Performance Surfaces (HPS) to improve recovery and reproducibility.
  • Provides charge-based separation and supreme resolution for acidic glycans.
  • Recommended for UPLC or UHPLC systems, particularly for LC-MS methods.
  • MaxPeak High Performance Surfaces (HPS) reduce analyte surface interactions, improving recovery and reproducibility.
  • Utilizes Ethylene-Bridged Hybrid (BEH) particle technology and anion-exchange features.
  • Integrated with MS-grade mobile-phase concentrate and Sialylated Glycan performance test standard.
  • Offers a wide pH range of 2 to 10 for diverse analyte analysis.
Catalog No. NC2001473