Learn More
Ace Glass, Inc. 25mm ID 8" 35/20 epoxy coated flash chromatography column with porosity B filter disc.
SDP
Supplier:  Ace Glass, Inc. 587264
Encompass_Preferred
25mm I.D., 8 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints. Includes porosity B filter disc.
Catalog No. 50-972-506