Learn More
Ace Glass, Inc. 50mm ID 24" 50/30 epoxy coated flash chromatography column.
SDP
Supplier:  Ace Glass, Inc. 587227
Encompass_Preferred
50mm I.D., 24 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 50/30 joints.
Catalog No. 50-972-480