Learn More
Ace Glass, Inc. 64mm ID 10" 50/30 epoxy coated flash chromatography column.
SDP
Supplier:  Ace Glass, Inc. 587229
Encompass_Preferred
64mm I.D., 10 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 50/30 joints.
Catalog No. 50-972-482